Learn More
Aviva Systems Biology Corp ARNTL Antibody - N-terminal region (ARP36822_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36822_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ARNTL. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of mouse ARNTL. Alternate names: MOP3, Arnt3, Bmal1, BMAL1b, bHLHe5, bmal1b'ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep
Catalog No. 50-014-56277