Learn More
Aviva Systems Biology Corp ARNTL Antibody - N-terminal region (ARP32002_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32002_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ARNTL. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human ARNTL. Alternate names: TIC, JAP3, MOP3, BMAL1, PASD3, BMAL1c, bHLHe5ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep
Catalog No. 50-014-54415