Learn More
Chem Service Inc Arochlor Calibration Standards Mixture - 8082
SDP
Catalog No. NC1615846
Encompass_Preferred
This item has a minimum order quantity of 2 per supplier requirements
Mixture of Arochlor 1016 (12674-11-2) and Arochlor 1260 (11096-82-5) in n-Hexane (110-54-3). Melting point/freezing point -137.74 F (-94.3 C) estimated, Initial boiling point 155.66 F (68.7 C) estimated, Flash point -7.0 F (-21.7 C) estimated.
Catalog No. NC1615846 Supplier Chem Service Inc Supplier No. MCSM8082J41ML
This item has a minimum order quantity of 2 per supplier requirements
Mixture of Arochlor 1016 (12674-11-2) and Arochlor 1260 (11096-82-5) in n-Hexane (110-54-3). Melting point/freezing point -137.74 F (-94.3 C) estimated, Initial boiling point 155.66 F (68.7 C) estimated, Flash point -7.0 F (-21.7 C) estimated.