Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aroclor 1016 and 1260 Calibration Standard (QUICKshots™ Working Level Standards/For EPA CLP Methods)

Catalog No. USCLP2811
Encompass
Change view
Click to view available options
Quantity:
1 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
USCLP2811 1 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. USCLP2811 Supplier Agilent Technologies Supplier No. CLP2811
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Specifications

Chemical Name or Material Aroclor 1016 and 1260 Calibration Standard
Packaging Ampule
Name Note QUICKshots™ Working Level Standards
Quantity 1 x 1 mL
Concentration or Composition (by Analyte or Components) Each component in Isooctane:
Aroclor 1016    •100ng/mL
Aroclor 1260    •100ng/mL
Decachlorobiphenyl    •20ng/mL
2,4,5,6-Tetrachloro-m-xylene    •20ng/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.