Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aroclor 1016, SPEX CertiPrep™
SDP

Catalog No. PCB1016 Shop All Spex CertiPrep Products
Change view
Click to view available options
Quantity:
1 mL
Packaging:
2mL amber prescored glass ampule
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
PCB1016 1 mL 2mL amber prescored glass ampule
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PCB1016 Supplier SPEX CertiPrep Supplier No. PCB1016

missing translation for 'validMainframeMsg'

Includes

Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
Aroclor 101612674-11-2200μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Chemical Identifiers

CAS 12674-11-2 , 110-54-3
Molecular Formula C12H7Cl3
Molecular Weight (g/mol) 257.538
InChI Key RIBGNAJQTOXRDK-UHFFFAOYSA-N
Synonym aroclor 1016, 3,3',5-trichlorobiphenyl, 1,1'-biphenyl, 3,3',5-trichloro, pcb 36, 3,3',5-trichloro-1,1'-biphenyl, 3,3/',5-trichlorobiphenyl, arochlor 1016, chlorodiphenyl cl, ccris 8481, dsstox_cid_3883
PubChem CID 38037
IUPAC Name 1,3-dichloro-5-(3-chlorophenyl)benzene
SMILES C1=CC(=CC(=C1)Cl)C2=CC(=CC(=C2)Cl)Cl

Specifications

CAS Max % 99.98, 0.02
Packaging 2mL amber prescored glass ampule
Quantity 1 mL
Concentration or Composition (by Analyte or Components) 200μg/mL
Chemical Name or Material Aroclor 1016
Hazard Category Highly flammable liquid and vapor. May be fatal if swallowed and enters airways. Causes skin irritation. May cause drowsiness or dizziness. Suspected of damaging fertility or the unborn child. May cause damage to organs through prolonged or repeated exposure.
DOTInformation Nonhazardous
Recommended Storage Ambient

Product intended for laboratory use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.