Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aroclor 1232 Solution, 1000μg/mL in isooctane, Ultra Scientific

Catalog No. USEPA1302
Encompass
Change view
Click to view available options
Quantity:
1 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
USEPA1302 1 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. USEPA1302 Supplier Agilent Technologies Supplier No. EPA13021
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Specifications

Chemical Name or Material Aroclor 1221 Solution
CAS 540-84-1, 11104-28-2
CAS Max % 99.85, 0.14
Analyte or Component Names 2,2,4-trimethylpentane (isooctane)
Element 2,2,4-trimethylpentane
Solvent or Matrix Iso-octane
Quantity 1 x 1 mL
Concentration or Composition (by Analyte or Components) 1000μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.