Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aroclor 1254 (PCB 1254) (For EPA 500, 600, 8000/ Appendix IX, CLP Methods)

Catalog No. USPP3511
Encompass
Change view
Click to view available options
Quantity:
1 mL (in Hexane)
1 mL (in Methanol)
Packaging:
Ampule
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
US-PP-3511 1 mL (in Hexane) Ampule
US-PP-3501 1 mL (in Methanol) Ampule
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. US-PP-3511 Supplier Agilent Technologies Supplier No. PP3511
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Aroclor 1254 at 100μg/mL in methanol or hexane

Chemical Identifiers

CAS 11097-69-1
Molecular Formula C12H5Cl5
Molecular Weight (g/mol) 326.422
InChI Key AUGNBQPSMWGAJE-UHFFFAOYSA-N
PubChem CID 40470
IUPAC Name 1,2,3-trichloro-4-(2,3-dichlorophenyl)benzene
SMILES C1=CC(=C(C(=C1)Cl)Cl)C2=C(C(=C(C=C2)Cl)Cl)Cl

Specifications

Packaging Ampule
Methods For EPA 500, 600, 8000/ Appendix IX
Quantity 1 mL (in Hexane)
Chemical Name or Material Aroclor 1254 (PCB 1254)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.