Learn More
Chem Service Inc AROCLOR 126 SOL AMPULE 5ML
SDP
Catalog No. NC9694805
Encompass_Preferred
This item has a minimum order quantity of 2 per supplier requirements
Arochlor 1260 Solution, 5ml, Ampuled: Yes, Solution/Mixture Specs: 1000microgram/ml in Hexane
Catalog No. NC9694805 Supplier Chem Service Inc Supplier No. S11092J45ML
This item has a minimum order quantity of 2 per supplier requirements
Arochlor 1260 Solution, 5ml, Ampuled: Yes, Solution/Mixture Specs: 1000microgram/ml in Hexane