Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aroclor 1268 (PCB 1268) (For EPA 500, 600, 8000/ Appendix IX, CLP Methods)

Catalog No. USPP3811
Encompass
Change view
Click to view available options
Quantity:
1 mL (in Hexane)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
US-PP-3811 1 mL (in Hexane)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. US-PP-3811 Supplier Agilent Technologies Supplier No. PP3811
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Aroclor 1268 at 100μg/mL in methanol or hexane

Specifications

Packaging Ampule
CAS 11100-14-4
Methods For EPA 500, 600, 8000/ Appendix IX
Quantity 1 mL (in Hexane)
Chemical Name or Material Aroclor 1268 (PCB 1268)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.