Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aroclor Solution Kits (For EPA 500, 600, 8000/ Appendix IX, CLP Methods)

Catalog No. USRPCK1A
Encompass
Change view
Click to view available options
Quantity:
9 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
US-RPCK-1A 9 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. US-RPCK-1A Supplier Agilent Technologies Supplier No. RPCK1A
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Chemical Name or Material Aroclor Solution Kits
Packaging Ampules
Methods For EPA 500, 600, 8000/ Appendix IX
Quantity 9 x 1 mL
Concentration or Composition (by Analyte or Components) Each kit contains one 1mL ampule each:
Arochlor 1016
Arochlor 1221
Arochlor 1232
Arochlor 1242
Arochlor 1248
Arochlor 1254
Arochlor 1260
Arochlor 1262
Arochlor 1268
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.