Learn More
Cayman Chemical Aromadendrene, Purity: ≥80%, CAS: 489-39-4, 10 mg
SDP
Catalog No. 502947138
Encompass_Preferred
A sesquiterpene with diverse biological activities; active against various strains of MRSA and VRE (MICs = 0.25-1 mg/ml); inhibits tert-butyl hydroperoxide-induced formation of TBARS in isolated rat liver hepatocytes at 1 mM; inhibits the proliferation of HepG2 and PC3 cells (IC50s = 20 and 9.3 µM, respectively)
Catalog No. 50-294-7138 Supplier Cayman Chemical Supplier No. 3382910 MG
A sesquiterpene with diverse biological activities; active against various strains of MRSA and VRE (MICs = 0.25-1 mg/ml); inhibits tert-butyl hydroperoxide-induced formation of TBARS in isolated rat liver hepatocytes at 1 mM; inhibits the proliferation of HepG2 and PC3 cells (IC50s = 20 and 9.3 µM, respectively)