Learn More
PREDICT HEALTH Coulumn for quantification of aromatic aminoacids in LCMS (1000 assays) columns
Supplier:  PREDICT HEALTH 140010AC
Encompass_Preferred
Intended Use: Monitoring of dietary treatment of PKU Disorder: PhenylketonuriaAnalytes: Phenylalanine, Tyrosine, TryptophanAnalysis Method: HPLC with UV detectorEvaluation: QuantitativeSample Type: Plasma and Serum
Catalog No. NC2891061