Learn More
ATCC Aromatoleum toluclasticum; Strain: TC-10
Catalog No. 502382216
Encompass
Aromatoleum toluclasticum strain TC-10 was isolated from creek water sediment from Tunitas creek. This strain is cited to be a dissimilatory iodate-reducing microorganism.
Catalog No. 50-238-2216 Supplier ATCC Supplier No. BAA3203
Aromatoleum toluclasticum strain TC-10 was isolated from creek water sediment from Tunitas creek. This strain is cited to be a dissimilatory iodate-reducing microorganism.