Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Arotinolol | 68377-92-4 | 99.9% | 10 MG
SDP

Catalog No. 5000230196
Encompass_Preferred

Arotinolol (CAS 68377-92-4) is a nonselective alpha and beta adrenergic receptor blocker used in research to study cardiovascular pharmacology and receptor binding. It is supplied as a research-grade chemical with defined purity and chemical identifiers.

  • Nonselective alpha/beta-adrenergic receptor blocker
  • Used in cardiovascular and receptor pharmacology research
  • Chemical formula C15H21N3O2S3 and molecular weight 371.54 g/mol
  • Reported high purity of 99.93%
  • Provided as a 10 mg research-grade solid for analytical studies

Catalog No. 50-002-30196 Supplier Medchemexpress LLC Supplier No. HY122537A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Arotinolol (CAS 68377-92-4) is a nonselective alpha and beta adrenergic receptor blocker used in research to study cardiovascular pharmacology and receptor binding. It is supplied as a research-grade chemical with defined purity and chemical identifiers.

  • Nonselective alpha/beta-adrenergic receptor blocker
  • Used in cardiovascular and receptor pharmacology research
  • Chemical formula C15H21N3O2S3 and molecular weight 371.54 g/mol
  • Reported high purity of 99.93%
  • Provided as a 10 mg research-grade solid for analytical studies

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.