Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Arotinolol-d5 hydrochloride | 2514848-16-7 | 413.03 | C15H17D5ClN3O2S3 | 1 MG
SDP

Catalog No. 5000283938
Encompass_Preferred

Arotinolol-d5 hydrochloride is a deuterium-labeled hydrochloride salt of arotinolol supplied as a research and analytical reference standard. It is intended for use as an isotopically labeled internal standard in mass spectrometry, tracer studies, and analytical method development; not for human therapeutic use.

  • Deuterium-labeled (d5) isotopologue for analytical applications.
  • Hydrochloride salt form for improved stability and handling.
  • Molecular weight 413.03.
  • Chemical formula C15H17D5ClN3O2S3.
  • CAS number 2514848-16-7.
  • Available as a 1 mg quantity suitable for trace-level reference use.

Catalog No. 50-002-83938 Supplier Medchemexpress LLC Supplier No. HYW6539691MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Arotinolol-d5 hydrochloride is a deuterium-labeled hydrochloride salt of arotinolol supplied as a research and analytical reference standard. It is intended for use as an isotopically labeled internal standard in mass spectrometry, tracer studies, and analytical method development; not for human therapeutic use.

  • Deuterium-labeled (d5) isotopologue for analytical applications.
  • Hydrochloride salt form for improved stability and handling.
  • Molecular weight 413.03.
  • Chemical formula C15H17D5ClN3O2S3.
  • CAS number 2514848-16-7.
  • Available as a 1 mg quantity suitable for trace-level reference use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.