Learn More
Medchemexpress LLC HY-103444 10mg Medchemexpress, ARP-100 CAS:704888-90-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10344410MG
Encompass_Preferred
Medchemexpress, HY-103444 10mg ARP-100 CAS:704888-90-4 ARP-100 is a potent and selective matrix metalloproteinase MMP-2 inhibitor (IC50=12 nM). ARP-100 interacts with S1' pocket of MMP-2 and shows anti-invasive properties in an in vitro model of invasion on matrigel. ARP-100 shows the less inhibitory activity towards MMP-1 (>50 μM), MMP-3 (4.5 μM), MMP-7 (>50 μM), and MMP-9 (0.2 μM). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0671