Learn More
ECM Bioscience Arp2/3 Complex Antibody Sampler Kit, ECM Biosciences
Catalog No. 502068603
Encompass_Preferred
The Arp2/3 complex kit can be used to detect expression levels of Arp2, Arp3, and Arpc1b in many different species. In addition, phospho-specific antibodies are included to detect phosphorylation of Arp2 (Thr-237/Thr-238) and Arpc1b (Thr-21). Applications: WB, E, IP, ICC; Species Reactivity: Hu, Rt, Ms
Catalog No. 50-206-8603 Supplier ECM Bioscience Supplier No. AK6670
The Arp2/3 complex kit can be used to detect expression levels of Arp2, Arp3, and Arpc1b in many different species. In addition, phospho-specific antibodies are included to detect phosphorylation of Arp2 (Thr-237/Thr-238) and Arpc1b (Thr-21). Applications: WB, E, IP, ICC; Species Reactivity: Hu, Rt, Ms