Learn More
ECM Bioscience Arp2/3 Complex Regulation Antibody Sampler Kit, ECM Biosciences
Catalog No. 502068600
Encompass_Preferred
The Arp2/3 complex regulation antibody sampler kit can be used to examine phosphorylation of the Arp2/3 complex subunit Arp2, as well as the Arp2/3 complex regulators N-WASP/WASP and Coronin-1B. The kit contains antibodies to monitor total expression levels of each of these proteins relative to site-specific phosphorylation. Applications: WB, E, ICC; Species Reactivity: Hu, Rt, Ms
Catalog No. 50-206-8600 Supplier ECM Bioscience Supplier No. AK6390
The Arp2/3 complex regulation antibody sampler kit can be used to examine phosphorylation of the Arp2/3 complex subunit Arp2, as well as the Arp2/3 complex regulators N-WASP/WASP and Coronin-1B. The kit contains antibodies to monitor total expression levels of each of these proteins relative to site-specific phosphorylation. Applications: WB, E, ICC; Species Reactivity: Hu, Rt, Ms