Learn More
Aviva Systems Biology Corp ARPC1B Antibody - C-terminal region (ARP65734_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP65734_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ARPC1B. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C-terminal region of Human ARPC1B. Alternate names: ARC41, p40-ARC, p41-ARCApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rat
Catalog No. 50-288-554