Learn More
Aviva Systems Biology Corp ARPC3 Antibody - middle region (ARP52093_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52093_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ARPC3. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human ARPC3. Alternate names: ARC21, p21-ArcApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-285-634