Learn More
ATCC ARPE-19; Retinal Epithelium; Human (Homo sapiens)
Catalog No. 502382088
Encompass
ARPE-19 is a spontaneously arising retinal pigment epithelia (RPE) cell line derived from the normal eyes of a 19-year-old male who died from head trauma in a motor vehicle accident. This cell line is a suitable transfection host.
Catalog No. 50-238-2088 Supplier ATCC Supplier No. CRL2302
 Restricted Item
Add to Cart
Add to Cart
ARPE-19 is a spontaneously arising retinal pigment epithelia (RPE) cell line derived from the normal eyes of a 19-year-old male who died from head trauma in a motor vehicle accident. This cell line is a suitable transfection host.