Learn More
Novus Biologicals™ Arpin Recombinant Protein Antigen

Description
Specifications
Specifications
| Gene ID (Entrez) | 348110 |
| Species | Human |
| Purity | >80% by SDS-PAGE and Coomassie blue staining |
| Content And Storage | Store at -20°C. Avoid freeze-thaw cycles. |
| Formulation | PBS and 1M Urea, pH 7.4. |
| For Use With (Application) | Antibody Competition |
| Gene Symbol | C15ORF38 |
| Molecular Weight (g/mol) | 26 kDa |
| Quantity | 100 μl |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.