Learn More
Aviva Systems Biology Corp Arpp21 Antibody - N-terminal region (ARP41873_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41873_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Arpp21. It was validated on Western Blot. . Alternate names: 0710001E13Rik, AI853636, ARPP-21, D9Bwg1012e, TarppApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-284-960