Learn More
Medchemexpress LLC HY-112215 5mg Medchemexpress, ARQ 531 CAS:2095393-15-8 Purity:>98%
SDP
Catalog No. 501969063
Encompass_Preferred
Medchemexpress, HY-112215 5mg ARQ 531 CAS:2095393-15-8 ARQ 531 is a reversible non-covalent inhibitor of Bruton’s Tyrosine Kinase (BTK), with IC50s of 0.85 nM and 0.39 nM for WT-BTK and C481S-BTK, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9063 Supplier Medchemexpress LLC Supplier No. HY1122155MG
Medchemexpress, HY-112215 5mg ARQ 531 CAS:2095393-15-8 ARQ 531 is a reversible non-covalent inhibitor of Bruton’s Tyrosine Kinase (BTK), with IC50s of 0.85 nM and 0.39 nM for WT-BTK and C481S-BTK, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.