Learn More
Medchemexpress LLC ARQ 621 | 1095253-39-6 | 98.09% | 552.43 | 1 MG
SDP
Catalog No. 5000393990
Encompass_Preferred
ARQ 621 is an allosteric, potent, and selective inhibitor of Eg5, which is a microtubule-based ATPase motor protein involved in cell division. It exhibits anti-tumor activity and functions as a kinesin inhibitor. Additionally, ARQ 621 is a click chemistry reagent, possessing an Alkyne group that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • Allosteric, potent, and selective inhibitor of Eg5
  • Targets microtubule-based ATPase motor protein involved in cell division
  • Demonstrates anti-tumor activity
  • Functions as a kinesin inhibitor
  • Click chemistry reagent for copper-catalyzed azide-alkyne cycloaddition (CuAAc)
Catalog No. 50-003-93990 Supplier Medchemexpress LLC Supplier No. HY160621MG
ARQ 621 is an allosteric, potent, and selective inhibitor of Eg5, which is a microtubule-based ATPase motor protein involved in cell division. It exhibits anti-tumor activity and functions as a kinesin inhibitor. Additionally, ARQ 621 is a click chemistry reagent, possessing an Alkyne group that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • Allosteric, potent, and selective inhibitor of Eg5
  • Targets microtubule-based ATPase motor protein involved in cell division
  • Demonstrates anti-tumor activity
  • Functions as a kinesin inhibitor
  • Click chemistry reagent for copper-catalyzed azide-alkyne cycloaddition (CuAAc)