Learn More
Aviva Systems Biology Corp ARR3 Antibody - middle region (ARP59750_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59750_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ARR3. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human ARR3. Alternate names: ARRXApplicationWB. Species reactivity: Human
Catalog No. 50-286-808