Learn More
Electron Microscopy Sciences Paraffin TMA ArrayMold D, Blue, 2.0 mm diameter, 36 cores per mold Capacity
SDP
Catalog No. 501902148
Encompass_Preferred
The Arraymold is the simple tool for laboratories to perform Paraffin tissue microarrays (TMAs) w/ less time while maintaining excellent results. Process up to 150 specimens onto one single slide w/ very little time consuming, minimal technical training. The DVD instruction is accompanying w/ each kit, will step by step guide you to perform through the entire TMAs process. Arraymolds allow pathologist or researcher to view many different samples on the same single slide. Helpful in finding IHC markers for control or quality assurance from hundreds of samples. Staining markers will be more consistent when hundreds of samples are on one slide. Used less costly antibody.Arraymold kit is small enough to be stored in a drawer.Rubber arraymold can be reused many times without losing its flexibility. Generates 2 mm diameter sample core, 36 cores per mold. Kit: 10 Disposable 2mm sterile dermal needles, 2 Stylets, 1 Brass Plate, 2mm reusable rubber Arraymold, DVD.Color: BlueDiameter: 2.0 mm
Catalog No. 50-190-2148 Supplier Electron Microscopy Sciences Supplier No. 6913015
The Arraymold is the simple tool for laboratories to perform Paraffin tissue microarrays (TMAs) w/ less time while maintaining excellent results. Process up to 150 specimens onto one single slide w/ very little time consuming, minimal technical training. The DVD instruction is accompanying w/ each kit, will step by step guide you to perform through the entire TMAs process. Arraymolds allow pathologist or researcher to view many different samples on the same single slide. Helpful in finding IHC markers for control or quality assurance from hundreds of samples. Staining markers will be more consistent when hundreds of samples are on one slide. Used less costly antibody.Arraymold kit is small enough to be stored in a drawer.Rubber arraymold can be reused many times without losing its flexibility. Generates 2 mm diameter sample core, 36 cores per mold. Kit: 10 Disposable 2mm sterile dermal needles, 2 Stylets, 1 Brass Plate, 2mm reusable rubber Arraymold, DVD.Color: BlueDiameter: 2.0 mm