Learn More
Electron Microscopy Sciences Paraffin TMA Arraymold Kit 2 (B +C), 1.5 and 4.0 mm diameter
SDP
Catalog No. 501900479
Encompass_Preferred
The Arraymold is the simple tool for laboratories to perform Paraffin tissue microarrays (TMAs) with less time while maintaining excellent results. Process up to 150 specimens onto one single slide with very little time consuming, minimal technical training. The DVD instruction is accompanying with each kit, will step by step guide you to perform through the entire TMAs process.
Arraymolds allow pathologist or researcher to view many different samples on the same single slide.
Helpful in finding IHC markers for control or quality assurance from hundreds of samples.
Staining markers will be more consistent when hundreds of samples are on one slide.
Used less costly antibody.
Arraymold kit is small enough to be stored in a drawer.
Rubber arraymold can be reused many times without losing its flexibility. his package includes two Arraymold kits, Kit B and Kit C.
Diameter: 1.5 and 4.0 mm
Catalog No. 50-190-0479 Supplier Electron Microscopy Sciences Supplier No. 6913103
The Arraymold is the simple tool for laboratories to perform Paraffin tissue microarrays (TMAs) with less time while maintaining excellent results. Process up to 150 specimens onto one single slide with very little time consuming, minimal technical training. The DVD instruction is accompanying with each kit, will step by step guide you to perform through the entire TMAs process.
Arraymolds allow pathologist or researcher to view many different samples on the same single slide.
Helpful in finding IHC markers for control or quality assurance from hundreds of samples.
Staining markers will be more consistent when hundreds of samples are on one slide.
Used less costly antibody.
Arraymold kit is small enough to be stored in a drawer.
Rubber arraymold can be reused many times without losing its flexibility. his package includes two Arraymold kits, Kit B and Kit C.
Diameter: 1.5 and 4.0 mm