Learn More
Aviva Systems Biology Corp ARRB2 Antibody - middle region (ARP48007_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48007_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ARRB2. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human ARRB2. Alternate names: ARB2, ARR2, DKFZp686L0365, BARR2ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-59702