Learn More
Montana Molecular Borealis Arrestin GLP-1R Assay - 10mL
SDP
Catalog No. NC2600161
Encompass_Preferred
Measure arrestin signaling kinetics in real time with the live cell, fluorescent Borealis arrestin - GLP-1 Receptor (GLP1R) assay. Add the unmodified GLP-1 receptor and Borealis sensor to cells and measure robust signals the next day on an automated plate reader or fluorescence imaging system. This assay is genetically encoded and packaged in BacMam for efficient transduction in most cell types - including uniform and near 100% efficiency in HEK293 cells as well as expression in many primary or iPSC-derived cells. The kit includes the arrestin sensor BacMam, GLP1R BacMam, GRK5 BacMam, 500mM sodium butyrate in H2O, and a positive control receptor and agonist (AT1R BacMam and 5mM Angiotensin II in H2O). Borealis kits for several other receptors are also available, and larger-sized kits are available on request. Other Montana Molecular tools including the cADDis cAMP sensor can be used alongside this assay.
Catalog No. NC2600161 Supplier Montana Molecular Supplier No. D2020G010
Measure arrestin signaling kinetics in real time with the live cell, fluorescent Borealis arrestin - GLP-1 Receptor (GLP1R) assay. Add the unmodified GLP-1 receptor and Borealis sensor to cells and measure robust signals the next day on an automated plate reader or fluorescence imaging system. This assay is genetically encoded and packaged in BacMam for efficient transduction in most cell types - including uniform and near 100% efficiency in HEK293 cells as well as expression in many primary or iPSC-derived cells. The kit includes the arrestin sensor BacMam, GLP1R BacMam, GRK5 BacMam, 500mM sodium butyrate in H2O, and a positive control receptor and agonist (AT1R BacMam and 5mM Angiotensin II in H2O). Borealis kits for several other receptors are also available, and larger-sized kits are available on request. Other Montana Molecular tools including the cADDis cAMP sensor can be used alongside this assay.