Learn More
Montana Molecular Borealis Arrestin LPA1R Assay - 10mL
SDP
Supplier:  Montana Molecular D2110G010
Encompass_Preferred
Measure arrestin signaling kinetics in real time with the live cell, fluorescent Borealis arrestin - LPA1 receptor assay. Add the unmodified LPA1 receptor and Borealis sensor to cells and measure robust signals the next day on an automated plate reader or fluorescence imaging system. This assay is genetically encoded and packaged in BacMam for efficient transduction in most cell types - including uniform and near 100% efficiency in HEK293 cells as well as expression in many primary or iPSC-derived cells. The kit includes the arrestin sensor BacMam, LPA1R BacMam, 500mM sodium butyrate in H2O, and a positive control receptor and agonist (AT1R BacMam and 5mM Angiotensin II in H2O). Borealis kits for several other receptors are also available, and larger-sized kits are available on request. Other Montana Molecular tools including the cADDis cAMP and DAG sensors can be used alongside this assay.
Catalog No. NC2600173