Learn More
Sigma Aldrich Fine Chemicals Biosciences Arrow electric lab stirrer1EA
Catalog No. 502908917
Encompass
Totally enclosed non-ventilated motor seals out corrosive fumes and dust. With on/off switch circuit-breaker and variable speed control. Includes a 9/16 x 10 in. support rod and 3/8 diam. x 12 in. L SS shaft and propeller.
Catalog No. 50-290-8917 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z1902681EA
Totally enclosed non-ventilated motor seals out corrosive fumes and dust. With on/off switch circuit-breaker and variable speed control. Includes a 9/16 x 10 in. support rod and 3/8 diam. x 12 in. L SS shaft and propeller.