Learn More
Ace Glass, Inc. Overhead DC electric stir motor, 1/15hp, 0-2000rpm, general-purpose, 2.43in-lbs torque, 120v 60Hz (220v 50Hz available), UL, CSA & ETL rated, Arrow Model 1750
SDP
Catalog No. NC9896153
Encompass
Arrow variable speed, general purpose overhead DC stir motor is designed to handle light to medium duty applications. Stir motor features a totally enclosed, non-ventilated motor and control housing IDeal for use in dusty or corrosive atmospheres and adds protection from splashes and spills. Features a 9/16 x 10 long integral support rod and a 3/8 motor shaft. Motors speed is 1/15hp, 0-2000rpm. UL, CSA, and ETL rated. Arrow model number is 1750.
Catalog No. NC9896153 Supplier Ace Glass, Inc. Supplier No. 1354420
Arrow variable speed, general purpose overhead DC stir motor is designed to handle light to medium duty applications. Stir motor features a totally enclosed, non-ventilated motor and control housing IDeal for use in dusty or corrosive atmospheres and adds protection from splashes and spills. Features a 9/16 x 10 long integral support rod and a 3/8 motor shaft. Motors speed is 1/15hp, 0-2000rpm. UL, CSA, and ETL rated. Arrow model number is 1750.