Learn More
TELEFLEX LLC Arrow™ StimuQuik™ Stimulating PNB Needle, 22ga, 2.5cm, 2.0 mm pin, 25/case
Catalog No. NC3803323
Encompass
Insulated needle shaft with a stimulating tip allows for needle guidance using stimulationDesigned with an atraumatic B-bevel tip for balance between tactile feedback and penetrationCentimeter markings on needle shaft for visual confirmation of insertion depthErgonomic needle hub for precision gripExtended length of injection tubing and electrical lead connections designed to help reduce the risk of contamination to the sterile field
Catalog No. NC3803323 Supplier TELEFLEX LLC Supplier No. AB22025SS
Insulated needle shaft with a stimulating tip allows for needle guidance using stimulationDesigned with an atraumatic B-bevel tip for balance between tactile feedback and penetrationCentimeter markings on needle shaft for visual confirmation of insertion depthErgonomic needle hub for precision gripExtended length of injection tubing and electrical lead connections designed to help reduce the risk of contamination to the sterile field