Learn More
Medchemexpress LLC HY-10531 5mg Medchemexpress, ARRY-380 (analog ) CAS:937265-83-3 Purity:>98%
SDP
Catalog No. 501968268
Encompass_Preferred
Medchemexpress, HY-10531 5mg ARRY-380 (analog ) CAS:937265-83-3 ARRY-380 analog, an inhibitor of EGFR (ErbB1), is extracted from patent WO2015153959A2, compound 249. ARRY-380 is a potent, selective, ATP-competitive, orally active inhibitor of HER2. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8268 Supplier Medchemexpress LLC Supplier No. HY105315MG
Medchemexpress, HY-10531 5mg ARRY-380 (analog ) CAS:937265-83-3 ARRY-380 analog, an inhibitor of EGFR (ErbB1), is extracted from patent WO2015153959A2, compound 249. ARRY-380 is a potent, selective, ATP-competitive, orally active inhibitor of HER2. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.