Learn More
Medchemexpress LLC HY-U00416 5mg Medchemexpress, ARS-1323 CAS:1698024-73-5 Purity:>98%
SDP
Catalog No. 501934440
Encompass_Preferred
Medchemexpress, HY-U00416 5mg ARS-1323 CAS:1698024-73-5 ARS-1323, the racemate of ARS-1620, is a novel inhibitor of mutant K-ras G12C extracted from patent WO 2015054572 A1. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4440 Supplier Medchemexpress LLC Supplier No. HYU004165MG
Medchemexpress, HY-U00416 5mg ARS-1323 CAS:1698024-73-5 ARS-1323, the racemate of ARS-1620, is a novel inhibitor of mutant K-ras G12C extracted from patent WO 2015054572 A1. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.