Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ars-1323-alkyne | 2436544-27-1 | MFCD32067946 | 99.6% | 569.00 g/mol | C28H27ClF2N6O3 | 5 MG
SDP

Catalog No. 5000262065
Encompass_Preferred

ARS-1323-alkyne is a covalent inhibitor probe that selectively targets the Switch-II pocket (S-IIP) of KRAS G12C. It is used to visualize covalent modification and to quantify inhibitor binding and target occupancy in biochemical and cellular studies; for research use only.

  • Covalent binding to the KRAS G12C Switch-II pocket.
  • Alkyne handle enables click-chemistry labeling and detection.
  • High purity (99.6%) suitable for biochemical and cellular assays.
  • Soluble in DMSO (250 mg/mL) and formulatable for in vivo dosing protocols.
  • Recommended storage: solid at 4°C under nitrogen; store solutions frozen for stability.
  • Molecular formula C28H27ClF2N6O3; molecular weight 569.00 g/mol.

Catalog No. 50-002-62065 Supplier Medchemexpress LLC Supplier No. HY1285225MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ARS-1323-alkyne is a covalent inhibitor probe that selectively targets the Switch-II pocket (S-IIP) of KRAS G12C. It is used to visualize covalent modification and to quantify inhibitor binding and target occupancy in biochemical and cellular studies; for research use only.

  • Covalent binding to the KRAS G12C Switch-II pocket.
  • Alkyne handle enables click-chemistry labeling and detection.
  • High purity (99.6%) suitable for biochemical and cellular assays.
  • Soluble in DMSO (250 mg/mL) and formulatable for in vivo dosing protocols.
  • Recommended storage: solid at 4°C under nitrogen; store solutions frozen for stability.
  • Molecular formula C28H27ClF2N6O3; molecular weight 569.00 g/mol.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.