Learn More
Selleck Chemical LLC ARS-1323-E1856-5MG
SDP
Catalog No. 503138889
Encompass_Preferred
ARS-1323 is the racemic mixture containing ARS-1620 and a potent inhibitor of mutant KRAS G12C It can be used in research for biochemical binding screens as companion imaging drugs (CID) for KRAS G12C
Catalog No. 50-313-8889 Supplier Selleck Chemical LLC Supplier No. E18565MG
ARS-1323 is the racemic mixture containing ARS-1620 and a potent inhibitor of mutant KRAS G12C It can be used in research for biochemical binding screens as companion imaging drugs (CID) for KRAS G12C