Learn More
Selleck Chemical LLC ARS-1620 2mg 1698055-85-4
SDP
Supplier:  Selleck Chemical LLC S87072MG
Encompass_Preferred
ARS-1620 is a potent, orally bioavailable covalent inhibitor of KRASG12C and could achieve rapid and sustained in vivo target occupancy to induce tumor regression. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8038