Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ars-1630 | 1698055-86-5 | 430.84 | 50 MG
SDP

Catalog No. 5000447977
Encompass_Preferred

ARS-1630 is a less active enantiomer of ARS-1620, identified as a novel inhibitor of mutant K-ras G12C. It is intended for research use only.

  • Solid appearance
  • Color ranges from white to yellow
  • Soluble in DMSO at ≥ 100 mg/mL
  • Inhibits mutant K-ras G12C by targeting Cys-12
  • Requires active cycling of KRASG12C between active-GTP and inactive-GDP conformations

Catalog No. 50-004-47977 Supplier Medchemexpress LLC Supplier No. HYU0041750MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ARS-1630 is a less active enantiomer of ARS-1620, identified as a novel inhibitor of mutant K-ras G12C. It is intended for research use only.

  • Solid appearance
  • Color ranges from white to yellow
  • Soluble in DMSO at ≥ 100 mg/mL
  • Inhibits mutant K-ras G12C by targeting Cys-12
  • Requires active cycling of KRASG12C between active-GTP and inactive-GDP conformations

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.