Learn More
TARGETMOL CHEMICALS INC ARS-1630 5MG
SDP
Catalog No. 502577457
Encompass_Preferred
Also available in 1mg 10mg 25mg 50mg 100mg and bulk. Please contact Fisher for quotes. ARS-1630 is a mutant K-ras G12C inhibitor. It is a less active enantiomer of ARS-1620. purity: 97%
Catalog No. 50-257-7457 Supplier TARGETMOL CHEMICALS INC Supplier No. T103765MG
Also available in 1mg 10mg 25mg 50mg 100mg and bulk. Please contact Fisher for quotes. ARS-1630 is a mutant K-ras G12C inhibitor. It is a less active enantiomer of ARS-1620. purity: 97%