Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC ARS-853 | 1629268-00-3 | 98.4% | 432.94 | 100 MG
SDP

Catalog No. 5000403347
Encompass_Preferred

ARS-853 is a cell-active, selective, covalent KRAS G12C inhibitor that targets mutant KRAS-driven signaling. It works by binding to the GDP-bound oncoprotein, thereby preventing its activation.

  • Cell-active, selective, covalent KRAS G12C inhibitor
  • Inhibits mutant KRAS-driven signaling
  • Binds to GDP-bound oncoprotein
  • Prevents KRAS activation
  • IC50 of 2.5 μM
  • Molecular weight of 432.94
  • Purity of 98.4%
  • Appearance as a solid
  • Color is white to off-white

Catalog No. 50-004-03347 Supplier Medchemexpress LLC Supplier No. HY19706100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ARS-853 is a cell-active, selective, covalent KRAS G12C inhibitor that targets mutant KRAS-driven signaling. It works by binding to the GDP-bound oncoprotein, thereby preventing its activation.

  • Cell-active, selective, covalent KRAS G12C inhibitor
  • Inhibits mutant KRAS-driven signaling
  • Binds to GDP-bound oncoprotein
  • Prevents KRAS activation
  • IC50 of 2.5 μM
  • Molecular weight of 432.94
  • Purity of 98.4%
  • Appearance as a solid
  • Color is white to off-white

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.