Learn More
Medchemexpress LLC HY-19706 1mg Medchemexpress, ARS-853 CAS:1629268-00-3 Purity:>98%
SDP
Catalog No. 502029287
Encompass_Preferred
Medchemexpress, HY-19706 1mg ARS-853 CAS:1629268-00-3 ARS-853 is a cell-active, selective, covalent KRAS G12C inhibitor with an IC50 of 2.5 μM. ARS-853 inhibits mutant KRAS-driven signaling by binding to the GDP-bound oncoprotein and preventing activation[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9287 Supplier Medchemexpress LLC Supplier No. HY197061MG
Medchemexpress, HY-19706 1mg ARS-853 CAS:1629268-00-3 ARS-853 is a cell-active, selective, covalent KRAS G12C inhibitor with an IC50 of 2.5 μM. ARS-853 inhibits mutant KRAS-driven signaling by binding to the GDP-bound oncoprotein and preventing activation[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.