Learn More
TARGETMOL CHEMICALS INC ARS-853 5MG
SDP
Catalog No. 502576315
Encompass_Preferred
Also available in 1mg 10mg 25mg 50mg 100mg 500mg and bulk. Please contact Fisher for quotes. ARS-853 is an inhibitor of K-RASG12C(IC50 : 2.5 μM) a mutant form of K-RAS that accumulates in the active GTP-bound state in certain cancer cellspurity: 98%
Catalog No. 50-257-6315 Supplier TARGETMOL CHEMICALS INC Supplier No. T74145MG
Also available in 1mg 10mg 25mg 50mg 100mg 500mg and bulk. Please contact Fisher for quotes. ARS-853 is an inhibitor of K-RASG12C(IC50 : 2.5 μM) a mutant form of K-RAS that accumulates in the active GTP-bound state in certain cancer cellspurity: 98%