Learn More
Selleck Chemical LLC ARS-853
SDP
Catalog No. 5000000198
Encompass_Preferred
ARS-853 is a selective covalent KRAS(G12C) inhibitor that inhibits mutant KRAS-driven signaling by binding to the GDP-bound oncoprotein and preventing activation ARS-853 also induces apoptosis
Catalog No. 50-000-00198 Supplier Selleck Chemical LLC Supplier No. S815610MM/1ML
ARS-853 is a selective covalent KRAS(G12C) inhibitor that inhibits mutant KRAS-driven signaling by binding to the GDP-bound oncoprotein and preventing activation ARS-853 also induces apoptosis