Learn More
Selleck Chemical LLC ARS-853
SDP
Supplier:  Selleck Chemical LLC S81565MG
Encompass_Preferred
ARS-853 is a selective covalent KRAS(G12C) inhibitor that inhibits mutant KRAS-driven signaling by binding to the GDP-bound oncoprotein and preventing activation ARS-853 also induces apoptosis
Catalog No. 50-000-00453