Learn More
Felix Storch Inc ACCUCOLD 18 Cu.Ft. Upright Vaccine Refrigerator, Certified to NSF/ANSI 456 Vaccine Storage Standard
SDP
Catalog No. NC2260536
Encompass_Preferred
ARS18PV456 18CF SOLID DOOR IS CERTIFIED PURPSOE BUILT for SAFE VACCINE STORAGE. Accucold is proud to lead the industry with the largest line of cold storage product sizes certified to NSF/ANSI 456 Vaccine Storage Standard. The Pharma-Vac Performance Series supports meeting regulations and guidelines from the CDC, State Agencies, World Health Organization, and other Health Ministries on vaccine storage and handling. Temp. Security and Saftey Features: https://www.accucold.com/product/ARS18PV456
Catalog No. NC2260536 Supplier Felix Storch Inc Supplier No. ARS18PV456
Add to Cart
Add to Cart
Request a Quote
ARS18PV456 18CF SOLID DOOR IS CERTIFIED PURPSOE BUILT for SAFE VACCINE STORAGE. Accucold is proud to lead the industry with the largest line of cold storage product sizes certified to NSF/ANSI 456 Vaccine Storage Standard. The Pharma-Vac Performance Series supports meeting regulations and guidelines from the CDC, State Agencies, World Health Organization, and other Health Ministries on vaccine storage and handling. Temp. Security and Saftey Features: https://www.accucold.com/product/ARS18PV456