Learn More
Felix Storch Inc ACCUCOLD 3 Cu.Ft. Counter Height Vaccine Refrigerator, Certified to NSF/ANSI 456 Vaccine Storage Standard
SDP
Catalog No. NC2260519
Encompass_Preferred
ARS3PV456 3CF SOLID DOOR IS CERTIFIED PURPSOE BUILT for SAFE VACCINE STORAGE. Accucold is proud to lead the industry with the largest line of cold storage product sizes certified to NSF/ANSI 456 Vaccine Storage Standard. The Pharma-Vac Performance Series supports meeting regulations and guidelines from the CDC, State Agencies, World Health Organization, and other Health Ministries on vaccine storage and handling. Temp. Security and Saftey Features: https://www.accucold.com/product/ARS3PV456
Catalog No. NC2260519 Supplier Felix Storch Inc Supplier No. ARS3PV456
ARS3PV456 3CF SOLID DOOR IS CERTIFIED PURPSOE BUILT for SAFE VACCINE STORAGE. Accucold is proud to lead the industry with the largest line of cold storage product sizes certified to NSF/ANSI 456 Vaccine Storage Standard. The Pharma-Vac Performance Series supports meeting regulations and guidelines from the CDC, State Agencies, World Health Organization, and other Health Ministries on vaccine storage and handling. Temp. Security and Saftey Features: https://www.accucold.com/product/ARS3PV456