Learn More
Felix Storch Inc ACCUCOLD ACCUCOLD ARS6PVDR Pharmacy-Vaccine Refrigerator 6 Cu. Ft. with Solid Door and Microprocessor control with +/- 1C variation
SDP
Catalog No. NC1859062
Encompass_Preferred
ACCUCOLD ARS6PVDR Pharmacy-Vaccine Refrigerator 6 Cu. Ft. with Solid Door with Advanced Temperature Control, cycle defrost and forced air cooling. Adjustable operating control range of 2C to 8C with Intelligent Microprocessor Digital Controller designed to provide +/- 1C variation. Multiple Audio and Visual Alarms for Safer Temperature Monitoring. Digital display of the min/max temperature (updated in 15 minute increments). Remote alarm contacts. LED lighting, adjustable shelves, 3/8' probe hole, reversible door. Meets EPA and SNAP requirements. Eco-Friendly using hydrocarbons for a greener global footprint. Compatible with Omnicell, AcuDose, and Pyxis (not included), ADA Option available. 2 Year Warranty Parts and Labor, 5-year compressor: https://www.accucold.com/product/ARS6PVDR
Catalog No. NC1859062 Supplier Felix Storch Inc Supplier No. ARS6PVDR
ACCUCOLD ARS6PVDR Pharmacy-Vaccine Refrigerator 6 Cu. Ft. with Solid Door with Advanced Temperature Control, cycle defrost and forced air cooling. Adjustable operating control range of 2C to 8C with Intelligent Microprocessor Digital Controller designed to provide +/- 1C variation. Multiple Audio and Visual Alarms for Safer Temperature Monitoring. Digital display of the min/max temperature (updated in 15 minute increments). Remote alarm contacts. LED lighting, adjustable shelves, 3/8' probe hole, reversible door. Meets EPA and SNAP requirements. Eco-Friendly using hydrocarbons for a greener global footprint. Compatible with Omnicell, AcuDose, and Pyxis (not included), ADA Option available. 2 Year Warranty Parts and Labor, 5-year compressor: https://www.accucold.com/product/ARS6PVDR