Learn More
Aviva Systems Biology Corp Arsb Antibody - middle region (ARP58861_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58861_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Arsb. It was validated on Western Blot. . Alternate names: 1110007C02Rik, AI480648, As-1, As-1r, As-1s, As-1t, As1, As1-r, As1-s, As1-t, Asr-1, Ast-1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rat
Catalog No. 50-286-507